missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLR1D Protein
A DNA sequence encoding the Homo sapiens (Human) POLR1D, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8289-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001193488.1 | |
POLR1D Protein | |
Research Use Only | |
POLR1D | |
Recombinant Protein | |
E. coli | |
MEEDQELERK ISGLKTSMAE GERKTALEMV QAAGTDRHCV TFVLHEEDHT LGNSLRYMIM KNPEVEFCGY TTTHPSESKI NLRIQTRGTL PAVEPFQRGL NELMNVCQHV LDKFEASIKD YKDQKASRNE STF | |
Greater than 80% as determined by SDS-PAGE. |
51082 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |