missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLL Protein
A cDNA sequence encoding the POLL was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | POLL Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952489
|
enQuireBio™
QP13094-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932489
|
enQuireBio™
QP13094-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942489
|
enQuireBio™
QP13094-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
POLL Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The POLL solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 30% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MLMHHQKYLQ RFLGGKREKK QKEACSIPGI GKRMAEKIIE ILESGHLRKL DHISESVPVL ELFSNIWGAG TKTAQMWYQQ GFRSLEDIRS QASLTTQQAI GLKHYSDFLE RMPREEATEI EQTVQKAAQA FNSGLLCVAC GSYRRGKATC GDVDVLITHP DGRSHRGIFS RLLDSLRQEG FLTDDLVSQE ENGQQQKYLG VCRLPGPGRR HRRLDIIVVP YSEFACALLY FTGSAHFNRS MRALAKTKGM SLSEHALSTA VVRNTHGCKV GPGRVLPTPT EKDVFRLLGL PYREPAERDW | |
Greater than 90% as determined by SDS-PAGE. |