missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PLAC8 Protein

A cDNA sequence encoding the PLAC8 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13075-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15962429

  • £2978.71 / 1mg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PLAC8 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PLAC8 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0),0.4M urea and 10% glycerol.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMQAQAPV VVVTQPGVGP GPAPQNSNWQ TGMCDCFSDC GVCLCGTFCF PCLGCQVAAD MNECCLCGTS VAMRTLYRTR YGIPGSICDD YMATLCCPHC TLCQIKRDIN RRRAMRTF
Videos
SDS
Documents
Certificates
Special Offers

Special Offers