missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PL 7 Protein
A cDNA sequence encoding the PL 7 was constructed and used to recombinantly synthesize the protein.
£99.15 - £4295.00
Specifications
Name | PL 7 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15972409
|
enQuireBio™
QP13069-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15962409
|
enQuireBio™
QP13069-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952409
|
enQuireBio™
QP13069-1MG |
1 mg |
£4295.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PL 7 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PL-7 is supplied in 20mM HEPES buffer pH 8.0, 200mM sodium chloride, and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MFEEKASSPS GKMGGEEKPI GAGEEKQKEG GKKKNKEGSG DGGRAELNPW PEYIYTRLEM YNILKAEHDS ILAEKAEKDS KPIKVTLPDG KQVDAESWKT TPYQIACGIS QGLADNTVIA KVNNVVWDLD RPLEEDCTLE LLKFEDEEAQ AVYWHSSAHI MGEAMERVYG GCLCYGPPIE NGFYYDMYLE EGGVSSNDFS SLEALCKKII KEKQAFERLE VKKETLLAMF KYNKFKCRIL NEKVNTPTTT VYRCGPLIDL CRGPHVRHTG KIKALKIHKN SSTYWEGKAD METLQRIYGI SFPDPKMLKE WEKFQEEAKN RDHRKIGRDQ ELYFFHELSP GSCFFLPKGA YIYNALIEFI RSEYRKRGFQ EVVTPNIFNS RLWMTSGHWQ HYSENMFSFE VEKELFALKP MNCPGHCLMF DHRPRSWREL PLRLADFGVL HRNELSGALT GLTRVRRFQQ DDAHIFCAME QIEDEIKGCL DFLRTVYSVF GFSFKLNLST RPEKFLGDIE VWDQAEKQLE NSLNEFGEKW ELNSGDGAFY GPKIDIQIKD AIGRYHQCAT IQLDFQLPIR FNLTYVSHDG DDKKRPVIVH RAILGSVERM IAILTENYGG KWPFWLSPRQ VMVVPVGPTC DEYAQKVRQQ FHDAKFMADI DLDPGCTLNK KIRNAQLAQY NFILVVGEKE KISGTVNIRT RDNKVHGERT ISETIERLQQ LKEFRSKQAE EEF | |
Greater than 90% as determined by SDS-PAGE. |