missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PKLR / PKRL Protein

A DNA sequence encoding the Homo sapiens (Human) PKLR / PKRL, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP1281-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961609

  • £384.00 / 100µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000289.1
PKLR / PKRL Protein
Research Use Only
Pklr
Recombinant Protein
E. coli
MSIQENISSL QLRSWVSKSQ RDLAKSILIG APGGPAGYLR RASVAQLTQE LGTAFFQQQQ LPAAMADTFL EHLCLLDIDS EPVAARSTSI IATIGPASRS VERLKEMIKA GMNIARLNFS HGSHEYHAES IANVREAVES FAGSPLSYRP VAIALDTKGP EIRTGILQGG PESEVELVKG SQVLVTVDPA FRTRGNANTV WVDYPNIVRV VPVGGRIYID DGLISLVVQK IGPEGLVTQV ENGGVLGSRK GVNLPGAQVD LPGLSEQDVR DLRFGVEHGV DIVFASFVRK ASDVAAVRAA LGPEGHGIKI ISKIENHEGV KRFDEILEVS DGIMVARGDL GIEIPAEKVF LAQKMMIGRC NLAGKPVVCA TQMLESMITK PRPTRAETSD VANAVLDGAD CIMLSGETAK GNFPVEAVKM QHAIAREAEA AVYHRQLFEE LRRAAPLSRD PTEVTAIGAV EAAFKCCAAA IIVLTTTGRS AQLLSRYRPR AAVIAVTRSA QAARQVHLCR GVFPLLYREP PEAIWADDVD RRVQFGIESG KLRGFLRVGD LVIVVTGWRP GSGYTNIMRV LSIS
Greater than 90% as determined by SDS-PAGE.
5313
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers