missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Phylloquinone omega-hydroxylase CYP4F11 Protein

A DNA sequence encoding the Homo sapiens (Human) Phylloquinone omega-hydroxylase CYP4F11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8115-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002751

  • £369.88 / 100µg

Estimated Shipment
02-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_001122404.1
Phylloquinone omega-hydroxylase CYP4F11 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
57834
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CYP4F11
Recombinant Protein
E. coli
TYTFYDNCRR LQCFPQPPKQ NWFWGHQGLV TPTEEGMKTL TQLVTTYPQG FKLWLGPTFP LLILCHPDII RPITSASAAV APKDMIFYGF LKPWLGDGLL LSGGDKWSRH RRMLTPAFHF NILKPYMKIF NKSVNIMHDK WQRLASEGSA RLDMFEHISL MTLDSLQKCV FSFESNCQEK PSEYIAAILE LSAFVEKRNQ QILLHTDFLY YLTPDGQRFR RACHLVHDFT DAVIQERRCT LPTQGIDDFL KNKAKSKTLD FIDVLLLSKD EDGKELSDED IRAEADTFMF EGHDTTASGL SWVLYHLAKH PEYQEQCRQE VQELLKDREP IEIEWDDLAQ LPFLTMCIKE SLRLHPPVPV ISRCCTQDFV LPDGRVIPKG IVCLINIIGI HYNPTVWPDP EVYDPFRFDQ ENIKERSPLA FIPFSAGPRN CIGQAFAMAE MKVVLALTLL HFRILPTHTE PRRKPELILR AEGGLWLRVE PLGANSQ
Videos
Documents