missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PHPT1 / PHP14 Protein
A DNA sequence encoding the Homo sapiens (Human) PHPT1 / PHP14, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6491-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001129333.1 | |
PHPT1 / PHP14 Protein | |
Research Use Only | |
PHPT1 | |
Recombinant Protein | |
Yeast | |
MAVADLALIP DVDIDSDGVF KYVLIRVHSA PRSGAPAAES KEIVRGYKWA EYHADIYDKV SGDMQKQGCD CECLGGGRIS HQSQDKKIHV YGYSMAYGPA QHAISTEKIK AKYPDYEVTW ANDGY | |
Greater than 90% as determined by SDS-PAGE. |
29085 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |