missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Phosphoglucomutase-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Phosphoglucomutase-1, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001166289.1 |
---|---|
Gene ID (Entrez) | 5236 |
Name | Phosphoglucomutase-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003650
|
enQuireBio™
QP6486-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023650
|
enQuireBio™
QP6486-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013650
|
enQuireBio™
QP6486-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033650
|
enQuireBio™
QP6486-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043650
|
enQuireBio™
QP6486-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053650
|
enQuireBio™
QP6486-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001166289.1 | |
Phosphoglucomutase-1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5236 | |
Research Use Only | |
PGM1 | |
Recombinant Protein | |
E. coli | |
MVKIVTVKTQ AYQDQKPGTS GLRKRVKVFQ SSANYAENFI QSIISTVEPA QRQEATLVVG GDGRFYMKEA IQLIARIAAA NGIGRLVIGQ NGILSTPAVS CIIRKIKAIG GIILTASHNP GGPNGDFGIK FNISNGGPAP EAITDKIFQI SKTIEEYAVC PDLKVDLGVL GKQQFDLENK FKPFTVEIVD SVEAYATMLR SIFDFSALKE LLSGPNRLKI RIDAMHGVVG PYVKKILCEE LGAPANSAVN CVPLEDFGGH HPDPNLTYAA DLVETMKSGE HDFGAAFDGD GDRNMILGKH GFFVNPSDSV AVIAANIFSI PYFQQTGVRG FARSMPTSGA LDRVASATKI ALYETPTGWK FFGNLMDASK LSLCGEESFG TGSDHIREKD GLWAVLAWLS ILATRKQSVE DILKDHWQKY GRNFFTRYDY EEVEAEGANK MMKDLEALMF DRSFVGKQFS ANDKVYTVEK ADNFEYSDPV DGSISRNQGL RLIFTDGSRI VFRLSGTGSA GATIRLYIDS YEKDVAKINQ DPQVMLAPLI SIALKVSQLQ ERTGRTAPTV IT | |
Greater than 90% as determined by SDS-PAGE. |