missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PHOSPHO1 Protein

A DNA sequence encoding the Homo sapiens (Human) PHOSPHO1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP1275-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951419

  • £560.06 / 100µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001137276.1
PHOSPHO1 Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
162466
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PHOSPHO1
Recombinant Protein
E. coli
MSGCFPVSGL RCLSRDGRMA AQGAPRFLLT FDFDETIVDE NSDDSIVRAA PGQRLPESLR ATYREGFYNE YMQRVFKYLG EQGVRPRDLS AIYEAIPLSP GMSDLLQFVA KQGACFEVIL ISDANTFGVE SSLRAAGHHS LFRRILSNPS GPDARGLLAL RPFHTHSCAR CPANMCKHKV LSDYLRERAH DGVHFERLFY VGDGANDFCP MGLLAGGDVA FPRRGYPMHR LIQEAQKAEP SSFRASVVPW ETAADVRLHL QQVLKSC
Videos
SDS
Documents
Certificates
Special Offers

Special Offers