missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PDK / PDZ binding kinase / TOPK Protein

A DNA sequence encoding the Homo sapiens (Human) PDK / PDZ binding kinase / TOPK, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7845-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051131

  • £1527.00 / 1mg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001265874.1
PDK / PDZ binding kinase / TOPK Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
55872
Greater than 80% as determined by SDS-PAGE.
Research Use Only
PBK
Recombinant Protein
E. coli
MEGISNFKTP SKLSEKKKSV LCSTPTINIP ASPFMQKLGF GTGVNVYLMK RSPRGLSHSP WAVKKINPIC NDHYRSVYQK RLMDEAKILK SLHHPNIVGY RAFTEANDGS LCLAMEYGGE KSLNDLIEER YKASQDPFPA AIILKVALNM ARGLKYLHQE KKLLHGDIKS SNVVIKGDFE TIKICDVGVS LPLDENMTVT DPEACYIGTE PWKPKEAVEE NGVITDKADI FAFGLTLWEM MTLSIPHINL SNDDDDEDKT FDESDFDDEA YYAALGTRPP INMEELDESY QKVIELFSVC TNEDPKDRPS AAHIVEALET DV
Videos
SDS
Documents
Certificates