missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDCL3 Protein
A cDNA sequence encoding the PDCL3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12989-5ug
Additional Details : Weight : 0.01000kg
Specifications
PDCL3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMQDPNAD TEWNDILRKK GILPPKESLK ELEEEAEEEQ RILQQSVVKT YEDMTLEELE DHEDEFNEED ERAIEMYRRR RLAEWKATKL KNKFGEVLEI SGKDYVQEVT KAGEGLWVIL HLYKQGIPLC ALINQHLSGL ARKFPDVKFI KAISTTCIPN YPDRNLPTIF VYLEGDIKAQ FIGPLVFGGM NLTRDELEWK LSESGAIMTD LEENPKKPIE DVLLSSVRRS VLMKRDSDSE GD | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PDCL3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl and 10% glycerol. |