missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PCP4L1 Protein
A cDNA sequence encoding the PCP4L1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12980-5ug
Additional Details : Weight : 0.01000kg
Specifications
PCP4L1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSELNTK TSPATNQAAG QEEKGKAGNV KKAEEEEEID IDLTAPETEK AALAIQGKFR RFQKRKKDPS S | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PCP4L1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 20% glycerol. |