missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PCNA Protein

A DNA sequence encoding the Homo sapiens (Human) PCNA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6470-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003530

  • £369.88 / 100µg

Estimated Shipment
05-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_002583.1
PCNA Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
5111
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PCNA
Recombinant Protein
E. coli
MFEARLVQGS ILKKVLEALK DLINEACWDI SSSGVNLQSM DSSHVSLVQL TLRSEGFDTY RCDRNLAMGV NLTSMSKILK CAGNEDIITL RAEDNADTLA LVFEAPNQEK VSDYEMKLMD LDVEQLGIPE QEYSCVVKMP SGEFARICRD LSHIGDAVVI SCAKDGVKFS ASGELGNGNI KLSQTSNVDK EEEAVTIEMN EPVQLTFALR YLNFFTKATP LSSTVTLSMS ADVPLVVEYK IADMGHLKYY LAPKIEDEEG S
Videos
Documents