missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PARP2 Protein
A cDNA sequence encoding the PARP2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12964-1mg
Additional Details : Weight : 0.01000kg
Specifications
PARP2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMQLDLR VQELIKLICN VQAMEEMMME MKYNTKKAPL GKLTVAQIKA GYQSLKKIED CIRAGQHGRA LMEACNEFYT RIPHDFGLRT PPLIRTQKEL SEKIQLLEAL GDIEIAIKLV KTELQSPEHP LDQHYRNLHC ALRPLDHESY EFKVISQYLQ STHAPTHSDY TMTLLDLFEV EKDGEKEAFR EDLHNRMLLW HGSRMSNWVG ILSHGLRIAP PEAPITGYMF GKGIYFADMS SKSANYCFAS RLKNTGLLLL SEVALGQCNE LLEANPKAEG LLQGKHSTKG LGKMAPSSAH FVTLNGSTVP LGPASDTGIL NPDGYTLNYN EYIVYNPNQV RMRYLLKVQF NFLQLW | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PARP2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |