missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PAIP2 Protein
A cDNA sequence encoding the PAIP2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12956-5ug
Additional Details : Weight : 0.01000kg
Specifications
PAIP2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MKDPSRSSTS PSIINEDVII NGHSHEDDNP FAEYMWMENE EEFNRQIEEE LWEEEFIERC FQEMLEEEEE HEWFIPARDL PQTMDQIQDQ FNDLVISDGS SLEDLVVKSN LNPNAKEFVP GVKYGNI | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PAIP2 protein solution contains 20mM Tris-HCl, pH 8.0, 1mM DTT and 10% Glycerol. |