missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PAFAH1B3 Protein

A cDNA sequence encoding the PAFAH1B3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12953-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15952059

  • £99.15 / 5µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

5050
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSGEENP ASKPTPVQDV QGDGRWMSLH HRFVADSKDK EPEVVFIGDS LVQLMHQCEI WRELFSPLHA LNFGIGGDGT QHVLWRLENG ELEHIRPKIV VVWVGTNNHG HTAEQVTGGI KAIVQLVNER QPQARVVVLG LLPRGQHPNP LREKNRQVNE LVRAALAGHP RAHFLDADPG FVHSDGTISH HDMYDYLHLS RLGYTPVCRA LHSLLLRLLA QDQGQGAPLL EPAP
Greater than 95% as determined by SDS-PAGE.
PAFAH1B3 Protein
Research Use Only
PAFAH1B3
Human
His
The PAFAH1B3 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers