missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PAFAH1B3 Protein
A cDNA sequence encoding the PAFAH1B3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12953-1mg
Additional Details : Weight : 0.01000kg
Specifications
5050 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSGEENP ASKPTPVQDV QGDGRWMSLH HRFVADSKDK EPEVVFIGDS LVQLMHQCEI WRELFSPLHA LNFGIGGDGT QHVLWRLENG ELEHIRPKIV VVWVGTNNHG HTAEQVTGGI KAIVQLVNER QPQARVVVLG LLPRGQHPNP LREKNRQVNE LVRAALAGHP RAHFLDADPG FVHSDGTISH HDMYDYLHLS RLGYTPVCRA LHSLLLRLLA QDQGQGAPLL EPAP | |
Greater than 95% as determined by SDS-PAGE. |
PAFAH1B3 Protein | |
Research Use Only | |
PAFAH1B3 | |
Human | |
His | |
The PAFAH1B3 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl and 10% glycerol. |