missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human p38a/SAPK2 Protein

A cDNA sequence encoding the p38a/SAPK2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12947-1ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15952039

  • £151.37 / 1µg
Estimated Shipment: 14-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

p38a/SAPK2 Protein
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
p38/SAPK2 is supplied in 25mM Tris-HCl, 150mM NaCl, 1mM DTT, 50% glycerol, pH 8.5.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MSQERPTFYR QELNKTIWEV PERYQNLSPV GSGAYGSVCA AFDTKTGLRV AVKKLSRPFQ SIIHAKRTYR ELRLLKHMKH ENVIGLLDVF TPARSLEEFN DVYLVTHLMG ADLNNIVKCQ KLTDDHVQFL IYQILRGLKY IHSADIIHRD LKPSNLAVNE DCELKILDFG LARHTDDEMT GYVATRWYRA PEIMLNWMHY NQTVDIWSVG CIMAELLTGR TLFPGTDHID QLKLILRLVG TPGAELLKKI SSESARNYIQ SLTQMPKMNF ANVFIGANPL AVDLLEKMLV LDSDKRITAA QALAHAYFAQ YHDPDDEPVA DPYDQSFESR DLLIDEWKSL TYDEVISFVP PPLDQEEMES
Videos
SDS
Documents
Certificates
Special Offers

Special Offers