missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OXSR1 Protein
A cDNA sequence encoding the OXSR1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12944-1mg
Additional Details : Weight : 0.01000kg
Specifications
OXSR1 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The OXSR1 solution (0.5 mg/1ml) contains Phosphate Buffered Saline (pH 7.4), 30% glycerol and 1mM DTT. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSEDSSA LPWSINRDDY ELQEVIGSGA TAVVQAAYCA PKKEKVAIKR INLEKCQTSM DELLKEIQAM SQCHHPNIVS YYTSFVVKDE LWLVMKLLSG GSVLDIIKHI VAKGEHKSGV LDESTIATIL REVLEGLEYL HKNGQIHRDV KAGNILLGED GSVQIADFGV SAFLATGGDI TRNKVRKTFV GTPCWMAPEV MEQVRGYDFK ADIWSFGITA IELATGAAPY HKYPPMKVLM LTLQNDPPSL ETGVQDKEML KKYGKSFRKM ISLCLQKDPE KRPTAAELLR HKFFQKAKNK EFLQEKTLQR APTISERAKK VRRVPGSSGR LHKTEDGGWE WSDDEFDEES EEGKAAISQL RSPRVKESIS NSELFPTTDP VGTLLQVPEQ ISAHLPQPAG QIATQPTQVS LPPTAEPAKT AQALSSGSGS QETKIPISLV LRLRNSKKEL NDIRFEFTPG RDTAEGVSQE LISAGLVDGR DLVIVAANLQ KIVEEPQSNR SVTFKLASGV EGSDIPDDGK LIGFAQLSIS |