missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OT Protein
A cDNA sequence encoding the OT was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12937-100mg
Additional Details : Weight : 0.01000kg
Specifications
5020 | |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Leu-Gly-NH2 | |
Greater than 99.0% as determined by RP-HPLC. |
OT Protein | |
Research Use Only | |
OXT | |
Human | |
Untagged | |
The Oxytocin was lyophilized from a concentrated (1 mg/ml) solution with no additives. |