missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ORM1 Protein

A cDNA sequence encoding the ORM1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12931-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951989

  • £155.00 / 10µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

5004
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Human
QIPLCANLVP VPITNATLDQ ITGKWFYIAS AFRNEEYNKS VQEIQATFFY FTPNKTEDTI FLREYQTRQD QCIYNTTYLN VQRENGTISR YVGGQEHFAH LLILRDTKTY MLAFDVNDEK NWGLSVYADK PETTKEQLGE FYEALDCLRI PKSDVVYTDW KKDKCEPLEK QHEKERKQEE GES
Greater than 95.0% as determined by SDS-PAGE.
ORM1 Protein
Research Use Only
ORM1
Human
Untagged
ORM1 protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 20mM TRIS and 50mM NaCl, pH 7.5.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers