missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ORM1 Protein
A cDNA sequence encoding the ORM1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12931-100ug
Additional Details : Weight : 0.01000kg
Specifications
5004 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
ORM1 | |
Human | |
Untagged | |
ORM1 protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 20mM TRIS and 50mM NaCl, pH 7.5. |
ORM1 Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
QIPLCANLVP VPITNATLDQ ITGKWFYIAS AFRNEEYNKS VQEIQATFFY FTPNKTEDTI FLREYQTRQD QCIYNTTYLN VQRENGTISR YVGGQEHFAH LLILRDTKTY MLAFDVNDEK NWGLSVYADK PETTKEQLGE FYEALDCLRI PKSDVVYTDW KKDKCEPLEK QHEKERKQEE GES |