missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OGN / osteoglycin Protein
A DNA sequence encoding the Homo sapiens (Human) OGN / osteoglycin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6449-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_054776.1 | |
OGN / osteoglycin Protein | |
Research Use Only | |
OGN | |
Recombinant Protein | |
Yeast | |
PPTQQDSRII YDYGTDNFEE SIFSQDYEDK YLDGKNIKEK ETVIIPNEKS LQLQKDEAIT PLPPKKENDE MPTCLLCVCL SGSVYCEEVD IDAVPPLPKE SAYLYARFNK IKKLTAKDFA DIPNLRRLDF TGNLIEDIED GTFSKLSLLE ELSLAENQLL KLPVLPPKLT LFNAKYNKIK SRGIKANAFK KLNNLTFLYL DHNALESVPL NLPESLRVIH LQFNNIASIT DDTFCKANDT SYIRDRIEEI RLEGNPIVLG KHPNSFICLK RLPIGSYF | |
Greater than 90% as determined by SDS-PAGE. |
4969 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |