missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human OAS1 Protein

A cDNA sequence encoding the OAS1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12915-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961939

  • £151.37 / 10µg
Estimated Shipment: 24-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

OAS1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The OAS1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MMDLRNTPAK SLDKFIEDYL LPDTCFRMQI NHAIDIICGF LKERCFRGSS YPVCVSKVVK GGSSGKGTTL RGRSDADLVV FLSPLTTFQD QLNRRGEFIQ EIRRQLEACQ RERAFSVKFE VQAPRWGNPR ALSFVLSSLQ LGEGVEFDVL PAFDALGQLT GSYKPNPQIY VKLIEECTDL QKEGEFSTCF TELQRDFLKQ RPTKLKSLIR LVKHWYQNCK KKLGKLPPQY ALELLTVYAW ERGSMKTHFN TAQGFRTVLE LVINYQQLCI YWTKYYDFKN PIIEKYLRRQ LTKPRPVILD PADPTGNLGG GDPKGWRQLA QEAEAWLNYP CFKNWDGSPV SSWILLVRPP ASSLPFIPAP LHEA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers