missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NXPH1 Protein

A cDNA sequence encoding the NXPH1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12912-HIS-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15941929

  • £133.00 / 10µg

Estimated Shipment
27-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NXPH1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The NXPH1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSANLTNGG KSELLKSGSS KSTLKHIWTE SSKDLSISRL LSQTFRGKEN DTDLDLRYDT PEPYSEQDLW DWLRNSTDLQ EPRPRAKRRP IVKTGKFKKM FGWGDFHSNI KTVKLNLLIT GKIVDHGNGT FSVYFRHNST GQGNVSVSLV PPTKIVEFDL AQQTVIDAKD SKSFNCRIEY EKVDKATKNT LCNYDPSKTC YQEQTQSHVS WLCSKPFKVI CIYISFYSTD YKLVQKVCPD YNYHSDTPYF PSG
Videos
SDS
Documents