missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nucleolar protein 1

A DNA sequence encoding the Homo sapiens (Human) Nucleolar protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6427-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003240

  • £613.01 / 200µg

Estimated Shipment
05-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_001171986.1
Nucleolar protein 1
200 μg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
8996
Greater than 90% as determined by SDS-PAGE.
Research Use Only
NOL3
Recombinant Protein
E. coli
MGNAQERPSE TIDRERKRLV ETLQADSGLL LDALLARGVL TGPEYEALDA LPDAERRVRR LLLLVQGKGE AACQELLRCA QRTAGAPDPA WDWQHVGPGY RDRSYDPPCP GHWTPEAPGS GTTCPGLPRA SDPDEAGGPE GSEAVQSGTP EEPEPELEAE ASKEAEPEPE PEPELEPEAE AEPEPELEPE PDPEPEPDFE ERDESEDS
Videos
Documents