missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nucleolar GTP-binding protein 1

A DNA sequence encoding the Homo sapiens (Human) Nucleolar GTP-binding protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7561-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049390

  • £384.00 / 100µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_037417.1
Nucleolar GTP-binding protein 1
Research Use Only
GNL2
Recombinant Protein
E. coli
MVKPKYKGRS TINPSKASTN PDRVQGAGGQ NMRDRATIRR LNMYRQKERR NSRGKIIKPL QYQSTVASGT VARVEPNIKW FGNTRVIKQS SLQKFQEEMD TVMKDPYKVV MKQSKLPMSL LHDRIRPHNL KVHILDTESF ETTFGPKSQR KRPNLFASDM QSLIENAEMS TESYDQGKDR DLVTEDTGVR NEAQEEIYKK GQSKRIWGEL YKVIDSSDVV VQVLDARDPM GTRSPHIETY LKKEKPWKHL IFVLNKCDLV PTWATKRWVA VLSQDYPTLA FHASLTNPFG KGAFIQLLRQ FGKLHTDKKQ ISVGFIGYPN VGKSSVINTL RSKKVCNVAP IAGETKVWQY ITLMRRIFLI DCPGVVYPSE DSETDIVLKG VVQVEKIKSP EDHIGAVLER AKPEYISKTY KIDSWENAED FLEKLAFRTG KLLKGGEPDL QTVGKMVLND WQRGRIPFFV KPPNAEPLVA PQLLPSSSLE VVPEAAQNNP GEEVTETAGE GSESIIKEET EENSHCDANT EMQQILTRVR QNFGKINVVP QFSGDDLVPV EVSDLEEELE SFSDEEEEEQ EQQRDDAEES SSEPEEENVG NDTKAVIKAL DEKIAKYQKF LDKAKAKKFS AVRISKGLSE KIFAKPEEQR KTLEEDVDDR APSKKGKKRK AQREEEQEHS NKAPRALTSK ERRRAVRQQR PKKVGVRYYE THNVKNRNRN KKKTNDSEGQ KHKRKKFRQK Q
Greater than 90% as determined by SDS-PAGE.
29889
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers