missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nuclear RNA export factor 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Nuclear RNA export factor 3, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_071335.1 |
---|---|
Gene ID (Entrez) | 56000 |
Name | Nuclear RNA export factor 3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083340
|
enQuireBio™
QP6443-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003350
|
enQuireBio™
QP6443-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093340
|
enQuireBio™
QP6443-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013350
|
enQuireBio™
QP6443-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023350
|
enQuireBio™
QP6443-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033350
|
enQuireBio™
QP6443-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_071335.1 | |
Nuclear RNA export factor 3 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
56000 | |
Research Use Only | |
NXF3 | |
Recombinant Protein | |
E. coli | |
MSLPSGHTTG HTDQVVQRRA RCWDIYQRRF SSRSEPVNPG MHSSSHQQQD GDAAMHGAHM DSPVRYTPYT ISPYNRKGSF RKQDQTHVNM EREQKPPERR MEGNMPDGTL GSWFKITVPF GIKYNEKWLL NLIQNECSVP FVPVEFHYEN MHASFFVENA SIAYALKNVS GKIWDEDNEK ISIFVNPAGI PHFVHRELKS EKVEQIKLAM NQQCDVSQEA LDIQRLPFYP DMVNRDTKMA SNPRKCMAAS LDVHEENIPT VMSAGEMDKW KGIEPGEKCA DRSPVCTTFS DTSSNINSIL ELFPKLLCLD GQQSPRATLC GTEAHKRLPT CKGSFFGSEM LKNLVLQFLQ QYYLIYDSGD RQGLLSAYHD EACFSLSIPF NPEDSAPSSF CKFFKDSRNI KILKDPYLRG ELLKHTKLDI VDSLSALPKT QHDLSSFLVD MWYQTEWMLC FSVNGVFKEV EGQSQGSVLA FTRTFIATPG SSSSLCIVND KLFVRDTSHQ GTQSALFTLV PTAFSSSVPA FSQEQQKMLP S | |
Greater than 90% as determined by SDS-PAGE. |