missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NSL1 Protein
A cDNA sequence encoding the NSL1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12886-5ug
Additional Details : Weight : 0.01000kg
Specifications
NSL1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAGSPEL VVLDPPWDKE LAAGTESQAL VSATPREDFR VRCTSKRAVT EMLQLCGRFV QKLGDALPEE IREPALRDAQ WTFESAVQEN ISINGQAWQE ASDNCFMDSD IKVLEDQFDE IIVDIATKRK QYPRKILECV IKTIKAKQEI LKQYHPVVHP LDLKYDPDPA PHMENLKCRG ETVAKEISEA MKSLPALIEQ GEGFSQVLRM QPVIHLQRIH QEVFSSCHRK PDAKPENFIT QIETTPTETA SRKTSDMVLK RKQTKDCPQR KWYPLRPKKI NLDT | |
Greater than 85% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NSL1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |