missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NRG1 A Protein
A cDNA sequence encoding the NRG1 A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10797-20ug
Additional Details : Weight : 0.01000kg
Specifications
NRG1 A Protein | |
Research Use Only | |
The ED50 was determined by the dose-dependent stimulation of the proliferation of human MCF-7 cells is less than 40 ng/ml, corresponding to a specific activity of > 2.5M 104 units/mg. | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered solution in 20mM PBS, pH 6.0 and 150mM NaCl. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
SHLVKCAEKE KTFCVNGGEC FMVKDLSNPS RYLCKCQPGF TGARCTENVP MKVQNQEKAE ELYQK | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |