missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NREP Protein
A cDNA sequence encoding the NREP was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12879-10ug
Additional Details : Weight : 0.01000kg
Specifications
9315 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMVYYPEL FVWVSQEPFP NKDMEGRLPK GRLPVPKEVN RKKNDETNAA SLTPLGSSEL RSPRISYLHF F | |
Greater than 85.0% as determined by SDS-PAGE. |
NREP Protein | |
Research Use Only | |
NREP | |
Human | |
His | |
The NREP solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |