missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NME3 Protein
A cDNA sequence encoding the NME3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12856-20ug
Additional Details : Weight : 0.01000kg
Specifications
NME3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MERTFLAVKP DGVQRRLVGE IVRRFERKGF KLVALKLVQA SEELLREHYA ELRERPFYGR LVKYMASGPV VAMVWQGLDV VRTSRALIGA TNPADAPPGT IRGDFCIEVG KNLIHGSDSV ESARREIALW FRADELLCWE DSAGHWLYE | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NME3 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 2mM DTT and 50% glycerol. |