missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NKX3-1 Protein

A cDNA sequence encoding the NKX3-1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12852-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15941749

  • £2975.00 / 1mg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NKX3-1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMLRVPEP RPGEAKAEGA APPTPSKPLT SFLIQDILRD GAQRQGGRTS SQRQRDPEPE PEPEPEGGRS RAGAQNDQLS TGPRAAPEEA ETLAETEPER HLGSYLLDSE NTSGALPRLP QTPKQPQKRS RAAFSHTQVI ELERKFSHQK YLSAPERAHL AKNLKLTETQ VKIWFQNRRY KTKRKQLSSE LGDLEKHSSL PALKEEAFSR ASLVSVYNSY PYYPYLYCVG SWSPAFW
Greater than 85.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
NKX3-1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2M UREA and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers