missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NHP2 Protein
A cDNA sequence encoding the NHP2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12845-5ug
Additional Details : Weight : 0.01000kg
Specifications
NHP2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTKIKADPDG PEAQAEACSG ERTYQELLVN QNPIAQPLAS RRLTRKLYKC IKKAVKQKQI RRGVKEVQKF VNKGEKGIMV LAGDTLPIEV YCHLPVMCED RNLPYVYIPS KTDLGAAAGS KRPTCVIMVK PHEEYQEAYD ECLEEVQSLP LPL | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NHP2 solution (0.25 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |