missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NHEJ1 Protein

A cDNA sequence encoding the NHEJ1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12842-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15941719

  • £1320.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

79840
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGS MEELEQG LLMQPWAWLQ LAENSLLAKV FITKQGYALL VSDLQQVWHE QVDTSVVSQR AKELNKRLTA PPAAFLCHLD NLLRPLLKDA AHPSEATFSC DCVADALILR VRSELSGLPF YWNFHCMLAS PSLVSQHLIR PLMGMSLALQ CQVRELATLL HMKDLEIQDY QESGATLIRD RLKTEPFEEN SFLEQFMIEK LPEACSIGDG KPFVMNLQDL YMAVTTQ
Greater than 90% as determined by SDS-PAGE.
NHEJ1 Protein
Research Use Only
NHEJ1
Human
His
The NHEJ1 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers