missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDUFS3 Protein

A cDNA sequence encoding the NDUFS3 was constructed and used to recombinantly synthesize the protein.

£99.15 - £1320.00

Specifications

Name NDUFS3 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15951649
View Documents Promotion Details
enQuireBio™
QP12819-1UG
1 μg
£99.15
1µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15971649
View Documents Promotion Details
enQuireBio™
QP12819-5UG
5 μg
£155.00
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15961649
View Documents Promotion Details
enQuireBio™
QP12819-50UG
50 μg
£1320.00
50µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NDUFS3 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The NDUFS3 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MESAGADTRP TVRPRNDVAH KQLSAFGEYV AEILPKYVQQ VQVSCFNELE VCIHPDGVIP VLTFLRDHTN AQFKSLVDLT AVDVPTRQNR FEIVYNLLSL RFNSRIRVKT YTDELTPIES AVSVFKAANW YEREIWDMFG VFFANHPDLR RILTDYGFEG HPFRKDFPLS GYVELRYDDE VKRVVAEPVE LAQEFRKFDL NSPWEAFPVY RQPPESLKLE AGDKKPDAK
Greater than 85.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications