missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFAF4 Protein
A cDNA sequence encoding the NDUFAF4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12815-5ug
Additional Details : Weight : 0.01000kg
Specifications
NDUFAF4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGALVIR GIRNFNLENR AEREISKMKP SVAPRHPSTN SLLREQISLY PEVKGEIARK DEKLLSFLKD VYVDSKDPVS SLQVKAAETC QEPKEFRLPK DHHFDMINIK SIPKGKISIV EALTLLNNHK LFPETWTAEK IMQEYQLEQK DVNSLLKYFV TFEVEIFPPE DKKAIRSK | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NDUFAF4 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |