missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAPA Protein
A cDNA sequence encoding the NAPA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12793-5ug
Additional Details : Weight : 0.01000kg
Specifications
NAPA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDNSGKEAEA MALLAEAERK VKNSQSFFSG LFGGSSKIEE ACEIYARAAN MFKMAKNWSA AGNAFCQAAQ LHLQLQSKHD AATCFVDAGN AFKKADPQEA INCLMRAIEI YTDMGRFTIA AKHHISIAEI YETELVDIEK AIAHYEQSAD YYKGEESNSS ANKCLLKVAG YAALLEQYQK AIDIYEQVGT NAMDSPLLKY SAKDYFFKAA LCHFCIDMLN AKLAVQKYEE LFPAFSDSRE CKLMKKLLEAHEEQNVDSYT ESVKEYDSIS RLDQWLTTML LRIKKTIQGD EEDLR | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NAPA protein solution (1 mg/ml) contains 20mM Tris-HCl pH 7.5 and 10% glycerol. |