missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NANS Protein
A cDNA sequence encoding the NANS was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | NANS Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951549
|
enQuireBio™
QP12790-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941549
|
enQuireBio™
QP12790-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931549
|
enQuireBio™
QP12790-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NANS Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NANS solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPLELELCPG RWVGGQHPCF IIAEIGQNHQ GDLDVAKRMI RMAKECGADC AKFQKSELEF KFNRKALERP YTSKHSWGKT YGEHKRHLEF SHDQYRELQR YAEEVGIFFT ASGMDEMAVE FLHELNVPFF KVGSGDTNNF PYLEKTAKKG RPMVISSGMQ SMDTMKQVYQ IVKPLNPNFC FLQCTSAYPL QPEDVNLRVI SEYQKLFPDI PIGYSGHETG IAISVAAVAL GAKVLERHIT LDKTWKGSDH SASLEPGELA ELVRSVRLVE RALGSPTKQL LPCEMACNEK LGKSVVAKVK IPEGTILTMD MLTVKVGEPK GYPPEDIFNL VGKKVLVTVE EDDTIMEELV DNHGKKIKS | |
Greater than 95.0% as determined by SDS-PAGE. |