missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human N-acetylgalactosamine-6-sulfatase Protein

A DNA sequence encoding the Homo sapiens (Human) N-acetylgalactosamine-6-sulfatase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6066-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15969729

  • £1735.33 / 1mg
Estimated Shipment: 04-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000503.1
N-acetylgalactosamine-6-sulfatase Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
2588
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GALNS
Recombinant Protein
E. coli
APQPPNILLL LMDDMGWGDL GVYGEPSRET PNLDRMAAEG LLFPNFYSAN PLCSPSRAAL LTGRLPIRNG FYTTNAHARN AYTPQEIVGG IPDSEQLLPE LLKKAGYVSK IVGKWHLGHR PQFHPLKHGF DEWFGSPNCH FGPYDNKARP NIPVYRDWEM VGRYYEEFPI NLKTGEANLT QIYLQEALDF IKRQARHHPF FLYWAVDATH APVYASKPFL GTSQRGRYGD AVREIDDSIG KILELLQDLH VADNTFVFFT SDNGAALISA PEQGGSNGPF LCGKQTTFEG GMREPALAWW PGHVTAGQVS HQLGSIMDLF TTSLALAGLT PPSDRAIDGL NLLPTLLQGR LMDRPIFYYR GDTLMAATLG QHKAHFWTWT NSWENFRQGI DFCPGQNVSG VTTHNLEDHT KLPLIFHLGR DPGERFPLSF ASAEYQEALS RITSVVQQHQ EALVPAQPQL NVCNWAVMNW APPGCEKLGK CLTPPESIPK KCLWSH
Videos
SDS
Documents
Certificates
Special Offers

Special Offers