missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human N-acetylgalactosamine-6-sulfatase Protein
A DNA sequence encoding the Homo sapiens (Human) N-acetylgalactosamine-6-sulfatase, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Accession Number | NP_000503.1 |
---|---|
Gene ID (Entrez) | 2588 |
Name | N-acetylgalactosamine-6-sulfatase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15909739
|
enQuireBio™
QP6066-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949739
|
enQuireBio™
QP6066-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919739
|
enQuireBio™
QP6066-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15939739
|
enQuireBio™
QP6066-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15959739
|
enQuireBio™
QP6066-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15929739
|
enQuireBio™
QP6066-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000503.1 | |
N-acetylgalactosamine-6-sulfatase Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2588 | |
Research Use Only | |
GALNS | |
Recombinant Protein | |
Yeast | |
APQPPNILLL LMDDMGWGDL GVYGEPSRET PNLDRMAAEG LLFPNFYSAN PLCSPSRAAL LTGRLPIRNG FYTTNAHARN AYTPQEIVGG IPDSEQLLPE LLKKAGYVSK IVGKWHLGHR PQFHPLKHGF DEWFGSPNCH FGPYDNKARP NIPVYRDWEM VGRYYEEFPI NLKTGEANLT QIYLQEALDF IKRQARHHPF FLYWAVDATH APVYASKPFL GTSQRGRYGD AVREIDDSIG KILELLQDLH VADNTFVFFT SDNGAALISA PEQGGSNGPF LCGKQTTFEG GMREPALAWW PGHVTAGQVS HQLGSIMDLF TTSLALAGLT PPSDRAIDGL NLLPTLLQGR LMDRPIFYYR GDTLMAATLG QHKAHFWTWT NSWENFRQGI DFCPGQNVSG VTTHNLEDHT KLPLIFHLGR DPGERFPLSF ASAEYQEALS RITSVVQQHQ EALVPAQPQL NVCNWAVMNW APPGCEKLGK CLTPPESIPK KCLWSH | |
Greater than 90% as determined by SDS-PAGE. |