missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MUSTN1 Protein

A cDNA sequence encoding the MUSTN1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12755-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961439

  • £1320.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

389125
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSQAGAQ EAPIKKKRPP VKEEDLKGAR GNLTKNQEIK SKTYQVMREC EQAGSAAPSV FSRTRTGTET VFEKPKAGPT KSVFG
Greater than 80.0% as determined by SDS-PAGE.
MUSTN1 Protein
Research Use Only
MUSTN1
Human
His
MUSTN1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers