missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MPPED2 Protein
A cDNA sequence encoding the MPPED2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12723-10ug
Additional Details : Weight : 0.01000kg
Specifications
744 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAHGIPS QGKVTITVDE YSSNPTQAFT HYNINQSRFQ PPHVHMVDPI PYDTPKPAGH TRFVCISDTH SRTDGIQMPY GDILLHTGDF TELGLPSEVK KFNDWLGNLP YEYKIVIAGN HELTFDKEFM ADLVKQDYYR FPSVSKLKPE DFDNVQSLLT NSIYLQDSEV TVKGFRIYGA PWTPWFNGWG FNLPRGQSLL DKWNLIPEGI DILMTHGPPL GFRDWVPKEL QRVGCVELLN TVQRRVRPKL HVFGGIHEGY GIMTDGYTTY INASTCTVSF QPTNPPIIFD LPNPQGS | |
Greater than 85% as determined by SDS-PAGE. |
MPPED2 Protein | |
Research Use Only | |
MPPED2 | |
Human | |
His | |
MPPED2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |