missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MORC family CW-type zinc finger protein 1

A DNA sequence encoding the Homo sapiens (Human) MORC family CW-type zinc finger protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7595-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049630

  • £560.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_056173.1
MORC family CW-type zinc finger protein 1
Research Use Only
MORC3
Recombinant Protein
E. coli
MAAQPPRGIR LSALCPKFLH TNSTSHTWPF SAVAELIDNA YDPDVNAKQI WIDKTVINDH ICLTFTDNGN GMTSDKLHKM LSFGFSDKVT MNGHVPVGLY GNGFKSGSMR LGKDAIVFTK NGESMSVGLL SQTYLEVIKA EHVVVPIVAF NKHRQMINLA ESKASLAAIL EHSLFSTEQK LLAELDAIIG KKGTRIIIWN LRSYKNATEF DFEKDKYDIR IPEDLDEITG KKGYKKQERM DQIAPESDYS LRAYCSILYL KPRMQIILRG QKVKTQLVSK SLAYIERDVY
Greater than 90% as determined by SDS-PAGE.
23515
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers