missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MLX-interacting Protein

A DNA sequence encoding the Homo sapiens (Human) MLX-interacting protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8054-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002401

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_055753.3
MLX-interacting Protein
Research Use Only
MLXIP
Recombinant Protein
E. coli
MDEQGCEHTS RTEDPFIQPT DFGPSEPPLS VPPPISPVLP LVPPPATALN PPAPPTFHQP QKFAGVNKAP SVITHTASAT LTHDAPATTF SQSQGLVITT HHPAPSAAPC GLALSPVTRP PQPRLTFVHP KPVSLTGGRP KQPHKIVPAP KPEPVSLVLK NARIAPGEPG GETQCGAPPD PEGCFPIPKA FKLVTTTTTL VCTCMRTHIH LNETKVS
Greater than 90% as determined by SDS-PAGE.
22877
500 μg
Not determined.
Full Length of Isoform 5
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers