missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MLEC Protein
A cDNA sequence encoding the MLEC was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12700-10ug
Additional Details : Weight : 0.01000kg
Specifications
MLEC Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSPGLGVAG VAGAAGAGLP ESVIWAVNAG GEAHVDVHGI HFRKDPLEGR VGRASDYGMK LPILRSNPED QILYQTERYN EETFGYEVPI KEEGDYVLVL KFAEVYFAQS QQKVFDVRLN GHVVVKDLDI FDRVGHSTAH DEIIPMSIRK GKLSVQGEVS TFTGKLYIEF VKGYYDNPKV CALYIMAGTV DDVPKLQPHP GLEKKEEEEE EEEYDEGSNL KKQTNKNRVQ SGPRTPNPYA SDNS | |
Greater than 95% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MLEC solution contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |