missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human METTL11A Protein
A DNA sequence encoding the Homo sapiens (Human) METTL11A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8164-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001273725.1 | |
METTL11A Protein | |
Research Use Only | |
NTMT1 | |
Recombinant Protein | |
E. coli | |
MTSEVIEDEK QFYSKAKTYW KQIPPTVDGM LGGYGHISSI DINSSRKFLQ RFLREGPNKT GTSCALDCGA GIGRITKRLL LPLFREVDMV DITEDFLVQA KTYLGEEGKR VRNYFCCGLQ DFTPEPDSYD VIWIQWVIGH LTDQHLAEFL RRCKGSLRPN GIIVIKDNMA QEGVILDDVD SSVCRDLDVV RRIICSAGLS LLAEERQENL PDEIYHVYSF ALR | |
Greater than 90% as determined by SDS-PAGE. |
28989 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |