missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human METAP1 Protein
A cDNA sequence encoding the METAP1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | METAP1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961219
|
enQuireBio™
QP12684-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951219
|
enQuireBio™
QP12684-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941219
|
enQuireBio™
QP12684-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
METAP1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The METAP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 3mM DTT, 40% glycerol, 200mM NaCl and 0.1mM PMSF. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAVETR VCETDGCSSE AKLQCPTCIK LGIQGSYFCS QECFKGSWAT HKLLHKKAKD EKAKREVSSW TVEGDINTDP WAGYRYTGKL RPHYPLMPTR PVPSYIQRPD YADHPLGMSE SEQALKGTSQ IKLLSSEDIE GMRLVCRLAR EVLDVAAGMIKPGVTTEEID HAVHLACIAR NCYPSPLNYY NFPKSCCTSV NEVICHGIPD RRPLQEGDIV NVDITLYRNG YHGDLNETFF VGEVDDGARK LVQTTYECLM QAIDAVKPGV RYRELGNIIQ KHAQANGFSV VRSYCGHGIH KLFHTAPNVP HYAKNKAVGV MKSGHVFTIE PMICEGGWQDETWPDGWTAV TRDGKRSAQF EHTLLVTDTG CEILTRRLDS ARPHFMSQF | |
Greater than 90.0% as determined by SDS-PAGE. |