missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Metalloproteinase inhibitor 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Metalloproteinase inhibitor 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9879-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003247.1 | |
Metalloproteinase inhibitor 4 Protein | |
Research Use Only | |
TIMP4 | |
Recombinant Protein | |
Yeast | |
CSCAPAHPQQ HICHSALVIR AKISSEKVVP ASADPADTEK MLRYEIKQIK MFKGFEKVKD VQYIYTPFDS SLCGVKLEAN SQKQYLLTGQ VLSDGKVFIH LCNYIEPWED LSLVQRESLN HHYHLNCGCQ ITTCYTVPCT ISAPNECLWT DWLLERKLYG YQAQHYVCMK HVDGTCSWYR GHLPLRKEFV DIVQP | |
Greater than 90% as determined by SDS-PAGE. |
7079 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |