missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MCEE Protein

A cDNA sequence encoding the MCEE was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12659-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961139

  • £2975.00 / 1mg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

84693
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MQVTGSVWNL GRLNHVAIAV PDLEKAAAFY KNILGAQVSE AVPLPEHGVS VVFVNLGNTK MELLHPLGRD SPIAGFLQKN KAGGMHHICI EVDNINAAVM DLKKKKIRSL SEEVKIGAHG KPVIFLHPKD CGGVLVELEQ A
Greater than 90% as determined by SDS-PAGE.
MCEE Protein
Research Use Only
MCEE
Human
His
The MCEE protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 1mM DTT, 0.1mM PMSF and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers